Product Details

SNP ID
rs35881748
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:54204474 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATACTAAGGCCAGAGAGGTTCAGCT[C/T]CTTAAAATATTCATGAAAATCCGAA
Phenotype
MIM: 611843
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CYB5RL PubMed Links

Gene Details

Gene
CYB5RL
Gene Name
cytochrome b5 reductase like
There are no transcripts associated with this gene.

Gene
MRPL37
Gene Name
mitochondrial ribosomal protein L37
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016491.3 Intron NP_057575.2
XM_005270920.3 Intron XP_005270977.1

View Full Product Details