Product Details

SNP ID
rs35178513
Assay Type
Functionally tested
NCBI dbSNP Submissions
33
Location
Chr.1:32211611 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTGACTTAGGGAACATCTAAGCAC[C/T]GTTTGGTGAAAGGTGTAAACTGTCT
Phenotype
MIM: 610162 MIM: 603911
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCDC28B PubMed Links

Gene Details

Gene
CCDC28B
Gene Name
coiled-coil domain containing 28B
There are no transcripts associated with this gene.

Gene
DCDC2B
Gene Name
doublecortin domain containing 2B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001099434.1 Intron NP_001092904.1
XM_017000413.1 Intron XP_016855902.1
XM_017000414.1 Intron XP_016855903.1
Gene
EIF3I
Gene Name
eukaryotic translation initiation factor 3 subunit I
There are no transcripts associated with this gene.

Gene
IQCC
Gene Name
IQ motif containing C
There are no transcripts associated with this gene.

Gene
TMEM234
Gene Name
transmembrane protein 234
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019118.4 Intron NP_061991.3
XM_011541795.2 Intron XP_011540097.1
XM_017001812.1 Intron XP_016857301.1
XM_017001813.1 Intron XP_016857302.1
XM_017001814.1 Intron XP_016857303.1
XM_017001815.1 Intron XP_016857304.1
XM_017001816.1 Intron XP_016857305.1
XM_017001817.1 Intron XP_016857306.1
XM_017001818.1 Intron XP_016857307.1
XM_017001819.1 Intron XP_016857308.1
XM_017001820.1 Intron XP_016857309.1
XM_017001821.1 Intron XP_016857310.1
XM_017001822.1 Intron XP_016857311.1
XM_017001823.1 Intron XP_016857312.1
XM_017001824.1 Intron XP_016857313.1
XM_017001825.1 Intron XP_016857314.1
XM_017001826.1 Intron XP_016857315.1
XM_017001827.1 Intron XP_016857316.1
XM_017001828.1 Intron XP_016857317.1
XM_017001829.1 Intron XP_016857318.1
XM_017001830.1 Intron XP_016857319.1
XM_017001831.1 Intron XP_016857320.1

View Full Product Details