Product Details

SNP ID
rs28675329
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:56656987 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAATGGGGCAGAGGCAGCATTTGA[A/G]GAGATAATGGCAGAAAGTATTCTAA
Phenotype
MIM: 600497
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PRKAA2 PubMed Links

Gene Details

Gene
PRKAA2
Gene Name
protein kinase AMP-activated catalytic subunit alpha 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006252.3 1553 Intron NP_006243.2
XM_017001692.1 1553 UTR 5 XP_016857181.1
XM_017001693.1 1553 Intron XP_016857182.1
XM_017001694.1 1553 Intron XP_016857183.1

View Full Product Details