Product Details
- SNP ID
-
rs28529311
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
30
- Location
-
Chr.1:58483588 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGATTTAAGATTTCCTTATATCTA[C/G]TGAGTGTTAAGTAAATGTTTAAGTA
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
OMA1
PubMed Links
Gene Details
- Gene
- OMA1
- Gene Name
- OMA1 zinc metallopeptidase
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_145243.4 |
|
Intron |
|
|
NP_660286.1 |
View Full Product Details