Product Details

SNP ID
rs28701522
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:32550154 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCGGATCTCTTGAGCCCAGGAGT[A/T]CAAGACCAGCCTGGGCAATATATCG
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ZBTB8A PubMed Links

Gene Details

Gene
ZBTB8A
Gene Name
zinc finger and BTB domain containing 8A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040441.2 Intron NP_001035531.2
NM_001291496.1 Intron NP_001278425.1

View Full Product Details