Product Details

SNP ID
rs28595301
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:70417954 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATTTTAGGTTTGGCCTTTGCTTCA[A/G]GTTTAGCAGCCACTGTAACTATTAC
Phenotype
MIM: 607657
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CTH PubMed Links

Gene Details

Gene
CTH
Gene Name
cystathionine gamma-lyase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190463.1 466 Intron NP_001177392.1
NM_001902.5 466 Missense Mutation AGT,GGT S90G NP_001893.2
NM_153742.4 466 Missense Mutation AGT,GGT S90G NP_714964.2
XM_005270509.3 466 UTR 5 XP_005270566.1
XM_017000416.1 466 Intron XP_016855905.1

View Full Product Details