Product Details
- SNP ID
-
rs12721040
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:116820764 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGTGGTCTCACCTCCCACTGGACAT[A/G]TGTCCTCAAGTTCATACCAGAACTT
- Phenotype
-
MIM: 107690
MIM: 107720
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
APOA4
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs35211609] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- APOA4
- Gene Name
- apolipoprotein A4
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000482.3 |
1398 |
UTR 3 |
|
|
NP_000473.2 |
- Gene
- APOC3
- Gene Name
- apolipoprotein C3
There are no transcripts associated with this gene.
View Full Product Details