Product Details

SNP ID
rs17880328
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:65183001 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCCATCCTGGGTGGGACTCGGCT[A/G]AGCCAGATCCTGGATGAGGAGTAGG
Phenotype
MIM: 114220 MIM: 614030
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CAPN1 PubMed Links

Gene Details

Gene
CAPN1
Gene Name
calpain 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001198868.1 Intron NP_001185797.1
NM_001198869.1 Intron NP_001185798.1
NM_005186.3 Intron NP_005177.2
XM_006718698.1 Intron XP_006718761.1
XM_011545292.1 Intron XP_011543594.1
Gene
SPDYC
Gene Name
speedy/RINGO cell cycle regulator family member C
There are no transcripts associated with this gene.

View Full Product Details