Product Details

SNP ID
rs11550159
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:65855420 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCAGCGGTCCTTGACCTCCTCGTA[A/G]CAGTTTGCTTGCAATTCATGCTTGA
Phenotype
MIM: 601442 MIM: 606591
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CFL1 PubMed Links

Gene Details

Gene
CFL1
Gene Name
cofilin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005507.2 651 Silent Mutation TGC,TGT C139C NP_005498.1
Gene
MUS81
Gene Name
MUS81 structure-specific endonuclease subunit
There are no transcripts associated with this gene.

Gene
SNX32
Gene Name
sorting nexin 32
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152760.2 651 Intron NP_689973.2
XM_006718488.3 651 Intron XP_006718551.1
XM_017017470.1 651 Intron XP_016872959.1
XM_017017471.1 651 Intron XP_016872960.1
XM_017017472.1 651 Intron XP_016872961.1
XM_017017473.1 651 Intron XP_016872962.1

View Full Product Details