Product Details

SNP ID
rs35861822
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:63539381 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAGAGCCTCTGAAAGTGACAGCAG[A/G]GCTGAGGATCTCATGGGGCTAGCAG
Phenotype
MIM: 605092
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RARRES3 PubMed Links

Gene Details

Gene
RARRES3
Gene Name
retinoic acid receptor responder 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004585.4 Intron NP_004576.2

View Full Product Details