Product Details

SNP ID
rs3210778
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:64227126 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGACGTACCTGGTGGGCACCCTGA[A/G]CCTCAGGTGAGATTCCAGGCTGGGC
Phenotype
MIM: 604189 MIM: 607901 MIM: 610470 MIM: 601398
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
DNAJC4 PubMed Links

Gene Details

Gene
DNAJC4
Gene Name
DnaJ heat shock protein family (Hsp40) member C4
There are no transcripts associated with this gene.

Gene
FERMT3
Gene Name
fermitin family member 3
There are no transcripts associated with this gene.

Gene
NUDT22
Gene Name
nudix hydrolase 22
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128612.2 532 Silent Mutation GAA,GAG E158E NP_001122084.1
NM_001128613.2 532 Silent Mutation GAA,GAG E158E NP_001122085.1
NM_001271831.1 532 Silent Mutation GAA,GAG E158E NP_001258760.1
NM_032344.3 532 Silent Mutation GAA,GAG E158E NP_115720.1
Gene
TRPT1
Gene Name
tRNA phosphotransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033678.3 532 Intron NP_001028850.2
NM_001160389.1 532 Intron NP_001153861.1
NM_001160390.1 532 Intron NP_001153862.1
NM_001160392.1 532 Intron NP_001153864.1
NM_001160393.1 532 Intron NP_001153865.1
NM_031472.3 532 Intron NP_113660.1
XM_005274345.4 532 Intron XP_005274402.1
XM_005274346.4 532 Intron XP_005274403.1
XM_005274347.4 532 Intron XP_005274404.1
XM_006718706.3 532 Intron XP_006718769.1
XM_006718707.3 532 Intron XP_006718770.1
XM_017018400.1 532 Intron XP_016873889.1
XM_017018401.1 532 Intron XP_016873890.1
Gene
VEGFB
Gene Name
vascular endothelial growth factor B
There are no transcripts associated with this gene.

View Full Product Details