Product Details

SNP ID
rs28663517
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:131181569 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATATTTTTGTTGCTTTAAATTTTT[C/T]AAGTGGAATAGTTATAAGAACAGCT
Phenotype
MIM: 615831
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LYRM7 PubMed Links

Gene Details

Gene
LYRM7
Gene Name
LYR motif containing 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001293735.1 Intron NP_001280664.1
NM_181705.3 Intron NP_859056.2

View Full Product Details