Product Details

SNP ID
rs35360938
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:138341396 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACTCTGGATGAGCTGAAATGCACA[C/T]GCTTCAGCATCAGTCTGGTAAAAGA
Phenotype
MIM: 157680 MIM: 609372
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CDC25C PubMed Links

Gene Details

Gene
CDC25C
Gene Name
cell division cycle 25C
There are no transcripts associated with this gene.

Gene
FAM53C
Gene Name
family with sequence similarity 53 member C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135647.1 506 Missense Mutation CGC,TGC R21C NP_001129119.1
NM_016605.2 506 Missense Mutation CGC,TGC R21C NP_057689.1

View Full Product Details