Product Details

SNP ID
rs34616588
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:130713922 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGAAAAGTTCTGGAGGAGTAGCCA[A/G]AGACCATCAGCGTTTCCTTTATGTG
Phenotype
MIM: 189980 MIM: 602238
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ABL1 PubMed Links

Gene Details

Gene
ABL1
Gene Name
ABL proto-oncogene 1, non-receptor tyrosine kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005157.5 42 Intron NP_005148.2
NM_007313.2 42 UTR 5 NP_009297.2
Gene
EXOSC2
Gene Name
exosome component 2
There are no transcripts associated with this gene.

View Full Product Details