Product Details

SNP ID
rs28407517
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:827993 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTGGGGCGTCAGAGGCGAAGGGCTC[C/T]GCAGGCTGGGACCTTGCCCCTGCAC
Phenotype
MIM: 602243 MIM: 614177 MIM: 609059
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CD151 PubMed Links

Gene Details

Gene
CD151
Gene Name
CD151 molecule (Raph blood group)
There are no transcripts associated with this gene.

Gene
CRACR2B
Gene Name
calcium release activated channel regulator 2B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286606.1 78 Intron NP_001273535.1
NM_173584.4 78 Intron NP_775855.3
XM_011520011.1 78 UTR 5 XP_011518313.1
XM_011520019.1 78 UTR 5 XP_011518321.1
XM_017017581.1 78 UTR 5 XP_016873070.1
XM_017017582.1 78 Intron XP_016873071.1
XM_017017583.1 78 Intron XP_016873072.1
XM_017017584.1 78 Intron XP_016873073.1
XM_017017585.1 78 Intron XP_016873074.1
XM_017017586.1 78 Intron XP_016873075.1
XM_017017587.1 78 Intron XP_016873076.1
XM_017017588.1 78 Intron XP_016873077.1
XM_017017589.1 78 UTR 5 XP_016873078.1
XM_017017590.1 78 Intron XP_016873079.1
XM_017017591.1 78 Intron XP_016873080.1
XM_017017592.1 78 Intron XP_016873081.1
XM_017017593.1 78 Intron XP_016873082.1
XM_017017594.1 78 Intron XP_016873083.1
XM_017017595.1 78 Intron XP_016873084.1
XM_017017596.1 78 UTR 5 XP_016873085.1
XM_017017597.1 78 UTR 5 XP_016873086.1
XM_017017598.1 78 UTR 5 XP_016873087.1
XM_017017599.1 78 Intron XP_016873088.1
Gene
PNPLA2
Gene Name
patatin like phospholipase domain containing 2
There are no transcripts associated with this gene.

View Full Product Details