Product Details

SNP ID
rs34704315
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:840264 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGTGGGGTATCGGATACACACAC[A/G]CTGCGGCCAGGCATTACGCCAGCTC
Phenotype
MIM: 602243 MIM: 614177 MIM: 601189 MIM: 602644
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CD151 PubMed Links

Gene Details

Gene
CD151
Gene Name
CD151 molecule (Raph blood group)
There are no transcripts associated with this gene.

Gene
CRACR2B
Gene Name
calcium release activated channel regulator 2B
There are no transcripts associated with this gene.

Gene
POLR2L
Gene Name
RNA polymerase II subunit L
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021128.4 333 UTR 3 NP_066951.1
Gene
TSPAN4
Gene Name
tetraspanin 4
There are no transcripts associated with this gene.

View Full Product Details