Product Details

SNP ID
rs28472692
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:849577 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTTCATCCCCAGGCCCCGGCCCCG[C/G]CCTCCAGTCCCCTCAGGCGCCCTTC
Phenotype
MIM: 601189 MIM: 602644
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
POLR2L PubMed Links

Gene Details

Gene
POLR2L
Gene Name
RNA polymerase II subunit L
There are no transcripts associated with this gene.

Gene
TSPAN4
Gene Name
tetraspanin 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001025234.1 Intron NP_001020405.1
NM_001025235.1 Intron NP_001020406.1
NM_001025236.1 Intron NP_001020407.1
NM_001025237.1 Intron NP_001020408.1
NM_001025238.1 Intron NP_001020409.1
NM_001025239.1 Intron NP_001020410.1
NM_003271.4 Intron NP_003262.1
XM_005253102.1 Intron XP_005253159.1
XM_005253104.1 Intron XP_005253161.1
XM_011520336.1 Intron XP_011518638.1
XM_011520337.2 Intron XP_011518639.1
XM_011520338.1 Intron XP_011518640.1
XM_011520339.2 Intron XP_011518641.1
XM_011520340.2 Intron XP_011518642.1
XM_011520341.2 Intron XP_011518643.1

View Full Product Details