Product Details

SNP ID
rs5814942
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:99567665 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGTGTGTGTGTGTGTGTGTGTGT[A/G]TTTTTGGAGAGCTCAGTCTCCATTT
Phenotype
MIM: 600660
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MEF2A PubMed Links

Gene Details

Gene
MEF2A
Gene Name
myocyte enhancer factor 2A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130926.2 811 Intron NP_001124398.1
NM_001130927.2 811 Intron NP_001124399.1
NM_001130928.2 811 Intron NP_001124400.1
NM_001171894.2 811 Intron NP_001165365.1
NM_001319206.1 811 Intron NP_001306135.1
NM_005587.3 811 Intron NP_005578.2
XM_005254914.2 811 Intron XP_005254971.1
XM_005254915.2 811 Intron XP_005254972.1
XM_005254916.3 811 Intron XP_005254973.1
XM_011521571.2 811 Intron XP_011519873.1
XM_011521572.2 811 Intron XP_011519874.1
XM_011521573.2 811 Intron XP_011519875.1
XM_011521576.2 811 Intron XP_011519878.1
XM_011521577.2 811 Intron XP_011519879.1
XM_011521578.2 811 Intron XP_011519880.1
XM_011521579.2 811 Intron XP_011519881.1
XM_011521581.2 811 Intron XP_011519883.1
XM_011521582.2 811 Intron XP_011519884.1
XM_011521583.2 811 Intron XP_011519885.1
XM_011521585.2 811 Intron XP_011519887.1
XM_011521586.2 811 Intron XP_011519888.1
XM_011521587.2 811 Intron XP_011519889.1
XM_011521590.2 811 Intron XP_011519892.1
XM_017022190.1 811 Intron XP_016877679.1
XM_017022191.1 811 Intron XP_016877680.1
XM_017022192.1 811 Intron XP_016877681.1
XM_017022193.1 811 Intron XP_016877682.1
XM_017022194.1 811 Intron XP_016877683.1
XM_017022195.1 811 Intron XP_016877684.1
XM_017022196.1 811 Intron XP_016877685.1
XM_017022197.1 811 UTR 5 XP_016877686.1
XM_017022198.1 811 Intron XP_016877687.1
XM_017022199.1 811 Intron XP_016877688.1
XM_017022200.1 811 Intron XP_016877689.1
XM_017022201.1 811 Intron XP_016877690.1
XM_017022202.1 811 Intron XP_016877691.1
XM_017022203.1 811 Intron XP_016877692.1
XM_017022204.1 811 Intron XP_016877693.1

View Full Product Details