Product Details
- SNP ID
-
rs28531997
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:248998 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TAGATATCTTTATTTTTATTGTGTA[C/T]GTAGAAGGAGTATTTGGCAAAACTC
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF876P
PubMed Links
Gene Details
- Gene
- ZNF876P
- Gene Name
- zinc finger protein 876, pseudogene
There are no transcripts associated with this gene.
View Full Product Details