Product Details

SNP ID
rs35860168
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:144510211 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCCCAGCTCTAGCCCCATCCCTGAG[C/T]GCTCTCTCGGGCTGCCCCACTTCTC
Phenotype
MIM: 138200 MIM: 609172 MIM: 603780
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GPT PubMed Links

Gene Details

Gene
GPT
Gene Name
glutamic--pyruvic transaminase
There are no transcripts associated with this gene.

Gene
LOC101928953
Gene Name
uncharacterized LOC101928953
There are no transcripts associated with this gene.

Gene
LRRC14
Gene Name
leucine rich repeat containing 14
There are no transcripts associated with this gene.

Gene
MFSD3
Gene Name
major facilitator superfamily domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138431.2 Intron NP_612440.1
XM_011516806.2 Intron XP_011515108.1
XM_017013005.1 Intron XP_016868494.1
Gene
PPP1R16A
Gene Name
protein phosphatase 1 regulatory subunit 16A
There are no transcripts associated with this gene.

Gene
RECQL4
Gene Name
RecQ like helicase 4
There are no transcripts associated with this gene.

View Full Product Details