Product Details
- SNP ID
-
rs1995285
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:20417738 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATAGACTAGGGCAATAAACACAGGC[A/G]AAAGTTGAGAATATGACTTATTTTT
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
HERC2P3
PubMed Links
Gene Details
- Gene
- HERC2P3
- Gene Name
- hect domain and RLD 2 pseudogene 3
There are no transcripts associated with this gene.
View Full Product Details