Product Details

SNP ID
rs3210514
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:1962215 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTTGGTGAATACAGTCTCCTTCCA[A/G]AGGTCGGGGGTCAGGTAGCTGTAGG
Phenotype
MIM: 603843 MIM: 603624 MIM: 605915
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NDUFB10 PubMed Links

Gene Details

Gene
NDUFB10
Gene Name
NADH:ubiquinone oxidoreductase subunit B10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004548.2 Intron NP_004539.1
Gene
RNF151
Gene Name
ring finger protein 151
There are no transcripts associated with this gene.

Gene
RPL3L
Gene Name
ribosomal protein L3 like
There are no transcripts associated with this gene.

Gene
RPS2
Gene Name
ribosomal protein S2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002952.3 Intron NP_002943.2
Gene
SNHG9
Gene Name
small nucleolar RNA host gene 9
There are no transcripts associated with this gene.

Gene
SNORA10
Gene Name
small nucleolar RNA, H/ACA box 10
There are no transcripts associated with this gene.

Gene
SNORA64
Gene Name
small nucleolar RNA, H/ACA box 64
There are no transcripts associated with this gene.

Gene
SNORA78
Gene Name
small nucleolar RNA, H/ACA box 78
There are no transcripts associated with this gene.

Gene
TBL3
Gene Name
transducin beta like 3
There are no transcripts associated with this gene.

View Full Product Details