Product Details

SNP ID
rs28811352
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:3498155 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCAGAAGTCCAAGAACCAGGAGCT[C/G]CAATGTCCAAGGGCACGGGAAGAAG
Phenotype
MIM: 616787
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
C16orf90 PubMed Links

Gene Details

Gene
C16orf90
Gene Name
chromosome 16 open reading frame 90
There are no transcripts associated with this gene.

Gene
CLUAP1
Gene Name
clusterin associated protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015041.2 Intron NP_055856.1
NM_024793.2 Intron NP_079069.1
XM_006720867.3 Intron XP_006720930.1
XM_017023068.1 Intron XP_016878557.1
XM_017023069.1 Intron XP_016878558.1
XM_017023070.1 Intron XP_016878559.1
XM_017023071.1 Intron XP_016878560.1
XM_017023072.1 Intron XP_016878561.1
XM_017023073.1 Intron XP_016878562.1

View Full Product Details