Product Details

SNP ID
rs17881650
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:4482186 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTCAGCAAAATTTTCAAATGTCTG[C/T]GTTCAGTGTCATATTTGTCCTGCTC
Phenotype
MIM: 141251
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HMOX2 PubMed Links

Gene Details

Gene
HMOX2
Gene Name
heme oxygenase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127204.1 Intron NP_001120676.1
NM_001127205.1 Intron NP_001120677.1
NM_001127206.2 Intron NP_001120678.1
NM_001286267.1 Intron NP_001273196.1
NM_001286268.1 Intron NP_001273197.1
NM_001286269.1 Intron NP_001273198.1
NM_001286270.1 Intron NP_001273199.1
NM_001286271.1 Intron NP_001273200.1
NM_002134.3 Intron NP_002125.3
XM_011522473.1 Intron XP_011520775.1
XM_017023196.1 Intron XP_016878685.1
XM_017023197.1 Intron XP_016878686.1
Gene
NMRAL1
Gene Name
NmrA-like family domain containing 1
There are no transcripts associated with this gene.

View Full Product Details