Product Details

SNP ID
rs28680478
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:536850 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGCAGGGTGGATTTCGGGAAGAG[C/T]ATCGGTGCCAGTTTCCCACAGTCAC
Phenotype
MIM: 603267
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CAPN15 PubMed Links

Gene Details

Gene
CAPN15
Gene Name
calpain 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005632.2 616 Intron NP_005623.1
XM_011522620.2 616 Intron XP_011520922.1
XM_011522621.2 616 UTR 5 XP_011520923.1
XM_011522622.1 616 Intron XP_011520924.1
XM_011522623.2 616 UTR 5 XP_011520925.1
XM_011522624.2 616 Intron XP_011520926.1
XM_011522625.2 616 Intron XP_011520927.1
XM_011522626.2 616 UTR 5 XP_011520928.1
XM_011522627.2 616 UTR 5 XP_011520929.1
XM_011522628.2 616 UTR 5 XP_011520930.1
XM_011522629.2 616 UTR 5 XP_011520931.1
XM_011522630.2 616 Intron XP_011520932.1
XM_011522631.2 616 Intron XP_011520933.1
XM_011522632.2 616 Intron XP_011520934.1
XM_017023596.1 616 UTR 5 XP_016879085.1
Gene
LINC00235
Gene Name
long intergenic non-protein coding RNA 235
There are no transcripts associated with this gene.

Gene
MIR3176
Gene Name
microRNA 3176
There are no transcripts associated with this gene.

Gene
MIR5587
Gene Name
microRNA 5587
There are no transcripts associated with this gene.

View Full Product Details