Product Details

SNP ID
rs11555899
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:12198747 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCAGAATGTGAAGGATTATTTACT[C/T]GATGATGGAACTCTGGTGGTTTCAG
Phenotype
MIM: 609230
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CDC123 PubMed Links

Gene Details

Gene
CDC123
Gene Name
cell division cycle 123
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006023.2 218 Silent Mutation CTC,CTT L39L NP_006014.2
XM_005252638.3 218 Silent Mutation CTC,CTT L39L XP_005252695.1
Gene
NUDT5
Gene Name
nudix hydrolase 5
There are no transcripts associated with this gene.

View Full Product Details