Product Details

SNP ID
rs28485673
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:83025356 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGGCAACAAAACAAGACTCCATC[A/C]CAAAAAAAAAAAAAAAAAAAAAAAG
Phenotype
MIM: 608530
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
BTBD1 PubMed Links

Gene Details

Gene
BTBD1
Gene Name
BTB domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001011885.1 Intron NP_001011885.1
NM_025238.3 Intron NP_079514.1

View Full Product Details