Product Details

SNP ID
rs28793737
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:22983802 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATGGTCAACCATAGTCTGAAAATA[T/G]GTGAGTATAGTACAGTGCTTATTTA
Phenotype
MIM: 606322 MIM: 608147
Polymorphism
T/G, Transversion substitution
Allele Nomenclature
Literature Links
CYFIP1 PubMed Links

Gene Details

Gene
CYFIP1
Gene Name
cytoplasmic FMR1 interacting protein 1
There are no transcripts associated with this gene.

Gene
TUBGCP5
Gene Name
tubulin gamma complex associated protein 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001102610.1 Intron NP_001096080.1
NM_052903.4 Intron NP_443135.3
XM_005272540.2 Intron XP_005272597.1
XM_011543868.1 Intron XP_011542170.1
XM_011543869.2 Intron XP_011542171.1
XM_011543870.1 Intron XP_011542172.1
XM_017021890.1 Intron XP_016877379.1
XM_017021891.1 Intron XP_016877380.1
XM_017021892.1 Intron XP_016877381.1
XM_017021893.1 Intron XP_016877382.1
XM_017021894.1 Intron XP_016877383.1
XM_017021895.1 Intron XP_016877384.1
XM_017021896.1 Intron XP_016877385.1
XM_017021897.1 Intron XP_016877386.1
XM_017021898.1 Intron XP_016877387.1
XM_017021899.1 Intron XP_016877388.1
XM_017021900.1 Intron XP_016877389.1
XM_017021901.1 Intron XP_016877390.1

View Full Product Details