Product Details

SNP ID
rs28595165
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:37420174 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAATATACAATAATTTTTGGCTCA[G/T]CGCAGTGGCTCACTCCTGTAATCAC
Phenotype
MIM: 602276
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TADA2A PubMed Links

Gene Details

Gene
TADA2A
Gene Name
transcriptional adaptor 2A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166105.2 Intron NP_001159577.2
NM_001291918.1 Intron NP_001278847.1
NM_001488.4 Intron NP_001479.4
NM_133439.3 Intron NP_597683.3
XM_006722044.3 Intron XP_006722107.1
XM_011525153.2 Intron XP_011523455.1
XM_011525154.2 Intron XP_011523456.1
XM_011525155.2 Intron XP_011523457.1
XM_017024981.1 Intron XP_016880470.1
XM_017024982.1 Intron XP_016880471.1
XM_017024983.1 Intron XP_016880472.1
XM_017024984.1 Intron XP_016880473.1
XM_017024985.1 Intron XP_016880474.1
XM_017024986.1 Intron XP_016880475.1

View Full Product Details