Product Details

SNP ID
rs28364626
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.17:36535494 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCGGCCGGAAGTGCCGCGCGGGAG[C/G]CGGAAGCGGCTGGTTCCCGCGTGGT
Phenotype
MIM: 612275 MIM: 610275
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
GGNBP2 PubMed Links

Gene Details

Gene
GGNBP2
Gene Name
gametogenetin binding protein 2
There are no transcripts associated with this gene.

Gene
MYO19
Gene Name
myosin XIX
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033580.2 491 Intron NP_001028752.1
NM_001163735.1 491 Intron NP_001157207.1
NM_025109.5 491 Intron NP_079385.2
XM_011525286.1 491 Intron XP_011523588.1
XM_011525287.1 491 Intron XP_011523589.1
XM_011525290.2 491 Intron XP_011523592.1
XM_017025157.1 491 Intron XP_016880646.1
XM_017025158.1 491 Intron XP_016880647.1
XM_017025159.1 491 Intron XP_016880648.1
XM_017025160.1 491 Intron XP_016880649.1
XM_017025161.1 491 Intron XP_016880650.1
Gene
PIGW
Gene Name
phosphatidylinositol glycan anchor biosynthesis class W
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178517.3 491 Intron NP_848612.2
XM_005257238.2 491 Intron XP_005257295.1
XM_011524646.2 491 UTR 5 XP_011522948.1

View Full Product Details