Product Details

SNP ID
rs28417559
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:36940410 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCACCTGCATGATGTGTAACAAGCA[A/G]CTCTCCACTGGCGAGGAACTCTACA
Phenotype
MIM: 608463 MIM: 601999
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AATF PubMed Links

Gene Details

Gene
AATF
Gene Name
apoptosis antagonizing transcription factor
There are no transcripts associated with this gene.

Gene
LHX1
Gene Name
LIM homeobox 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005568.4 1014 Silent Mutation CAA,CAG Q97Q NP_005559.2
Gene
LOC102723471
Gene Name
uncharacterized LOC102723471
There are no transcripts associated with this gene.

View Full Product Details