Product Details

SNP ID
rs28554117
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:48541777 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGACTTAGGATTTTTCAACTTTAC[A/G]ATGGTGCAAAAGCGATACGCATTTG
Phenotype
MIM: 311240
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TBC1D25 PubMed Links

Gene Details

Gene
TBC1D25
Gene Name
TBC1 domain family member 25
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002536.2 Intron NP_002527.1

View Full Product Details