Product Details

SNP ID
rs11798830
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:48702684 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGAAGACTACACCCCAAGGGCAGC[C/G]TCAGACCCTCCTGGGCCTGAGACTT
Phenotype
MIM: 300254
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SUV39H1 PubMed Links

Gene Details

Gene
SUV39H1
Gene Name
suppressor of variegation 3-9 homolog 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282166.1 Intron NP_001269095.1
NM_003173.3 Intron NP_003164.1

View Full Product Details