Product Details

SNP ID
rs11538179
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:48835484 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCCGAGCAGCAGCAGCACCAAAA[C/G]GCCGACGCCCCCGGCCCGCGGCCCC
Phenotype
MIM: 300437 MIM: 300399
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ERAS PubMed Links

Gene Details

Gene
ERAS
Gene Name
ES cell expressed Ras
There are no transcripts associated with this gene.

Gene
PCSK1N
Gene Name
proprotein convertase subtilisin/kexin type 1 inhibitor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013271.4 155 Missense Mutation CTT,GTT L17V NP_037403.1

View Full Product Details