Product Details

SNP ID
hCV64435365
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:48894819 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGTTCAGAGAGGTTAAGCCACTA[G/T]CCCAAGGTCACATGGCAGGGAACAG
Phenotype
MIM: 300463 MIM: 314375 MIM: 300249
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PQBP1 PubMed Links

Gene Details

Gene
PQBP1
Gene Name
polyglutamine binding protein 1
There are no transcripts associated with this gene.

Gene
SLC35A2
Gene Name
solute carrier family 35 member A2
There are no transcripts associated with this gene.

Gene
TIMM17B
Gene Name
translocase of inner mitochondrial membrane 17 homolog B (yeast)
There are no transcripts associated with this gene.

View Full Product Details