Product Details

SNP ID
rs6609049
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:40055051 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGACCTAACACCATAAACCTGTCCA[A/G]ATAATTTCATGTATGGAAGGTTTGA
Phenotype
MIM: 300485
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BCOR PubMed Links

Gene Details

Gene
BCOR
Gene Name
BCL6 corepressor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001123383.1 Intron NP_001116855.1
NM_001123384.1 Intron NP_001116856.1
NM_001123385.1 Intron NP_001116857.1
NM_017745.5 Intron NP_060215.4
XM_005272616.1 Intron XP_005272673.1
XM_005272618.3 Intron XP_005272675.1
XM_005272619.3 Intron XP_005272676.1
XM_005272620.3 Intron XP_005272677.1
XM_006724536.3 Intron XP_006724599.1
XM_011543929.2 Intron XP_011542231.1
XM_011543930.1 Intron XP_011542232.1
XM_011543931.2 Intron XP_011542233.1
XM_017029615.1 Intron XP_016885104.1
XM_017029616.1 Intron XP_016885105.1

View Full Product Details