Product Details
- SNP ID
-
rs28940593
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:37686111 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTTTCTCGCTACTGCCTTGCGCGCT[C/T]GTGCAGCTCACGCTTCTCTTCGTAC
- Phenotype
-
MIM: 314850
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
LANCL3
PubMed Links
Gene Details
- Gene
- LANCL3
- Gene Name
- LanC like 3
There are no transcripts associated with this gene.
- Gene
- XK
- Gene Name
- X-linked Kx blood group
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
XM_011543978.2 |
346 |
Silent Mutation |
CTC,CTT |
L50L |
XP_011542280.1 |
View Full Product Details