Product Details
- SNP ID
-
hCV64446854
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:43766615 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAAAAAGAAAGGACAGCAGATATGG[G/T]CAATCAACTTTTATTTTTAATTACC
- Phenotype
-
MIM: 309860
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
MAOB
PubMed Links
Gene Details
- Gene
- MAOB
- Gene Name
- monoamine oxidase B
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000898.4 |
2590 |
UTR 3 |
|
|
NP_000889.3 |
View Full Product Details