Product Details

SNP ID
rs61759298
Assay Type
Functionally tested
NCBI dbSNP Submissions
27
Location
Chr.1:3745187 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCCCAGGCTCCTTGGGCCTGGGA[C/T]CTCCTTGCCTAGCCCTCACCCCATT
Phenotype
MIM: 601990
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCDC27 PubMed Links

Gene Details

Gene
CCDC27
Gene Name
coiled-coil domain containing 27
There are no transcripts associated with this gene.

Gene
TP73
Gene Name
tumor protein p73
There are no transcripts associated with this gene.

Gene
TP73-AS1
Gene Name
TP73 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details