Product Details

SNP ID
rs61742168
Assay Type
Functionally tested
NCBI dbSNP Submissions
23
Location
Chr.1:153438301 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGAAACATCGCGACTATGTCCAAC[A/G]TGACTTTCTCACCTAGAGGGATATT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
S100A7L2 PubMed Links

Gene Details

Gene
S100A7L2
Gene Name
S100 calcium binding protein A7 like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001045479.1 140 Missense Mutation ACG,ATG T21M NP_001038944.2

View Full Product Details