Product Details

SNP ID
rs61736237
Assay Type
Functionally tested
NCBI dbSNP Submissions
9
Location
Chr.1:248387692 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAATAAATGCTCAGGATTCTTTTT[C/T]GGTGTCATTTGTGCCGTCTTCTTCA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR2T6 PubMed Links

Gene Details

Gene
OR2T6
Gene Name
olfactory receptor family 2 subfamily T member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005471.1 Intron NP_001005471.1

View Full Product Details