Product Details

SNP ID
rs41269351
Assay Type
Functionally tested
NCBI dbSNP Submissions
31
Location
Chr.1:248406373 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATTGACATGATGTATATTTCCACT[A/G]TTGTGCCTAAGATGCTGGTTAATTA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR2T1 PubMed Links

Gene Details

Gene
OR2T1
Gene Name
olfactory receptor family 2 subfamily T member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030904.1 379 Missense Mutation ATT,GTT I127V NP_112166.1

View Full Product Details