Product Details

SNP ID
rs61747448
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:207712007 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTCCGACATCTACGAGGCGGTGTC[C/T]GGGGACGCGGTGGCGGTAGCGCCCG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCNYL1 PubMed Links

Gene Details

Gene
CCNYL1
Gene Name
cyclin Y like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142300.1 216 Silent Mutation TCC,TCT S37S NP_001135772.1
NM_152523.2 216 UTR 5 NP_689736.1
XM_005246343.1 216 Intron XP_005246400.1

View Full Product Details