Product Details

SNP ID
rs148800637
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:218675404 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACTATCTGCATTCCCACCGCATCC[C/T]ACACCGAGATATGAAGCCTCAGAAC
Phenotype
MIM: 616014 MIM: 607652
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RNF25 PubMed Links

Gene Details

Gene
RNF25
Gene Name
ring finger protein 25
There are no transcripts associated with this gene.

Gene
STK36
Gene Name
serine/threonine kinase 36
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243313.1 312 Missense Mutation CCA,CTA P122L NP_001230242.1
NM_015690.4 312 Missense Mutation CCA,CTA P122L NP_056505.2
XM_005246464.1 312 Missense Mutation CCA,CTA P122L XP_005246521.1
XM_011510959.1 312 Missense Mutation CCA,CTA P122L XP_011509261.1
XM_011510960.1 312 Missense Mutation CCA,CTA P68L XP_011509262.1
XM_011510961.1 312 Intron XP_011509263.1
XM_017003804.1 312 Missense Mutation CCA,CTA P122L XP_016859293.1
XM_017003805.1 312 UTR 5 XP_016859294.1

View Full Product Details