Product Details

SNP ID
rs61730110
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:153162771 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCTCACCGTGCAGTTCGTGACGGG[C/T]TCCTTCATCGAGAAGTACGACCCGA
Phenotype
MIM: 179541
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RAP2B PubMed Links

Gene Details

Gene
RAP2B
Gene Name
RAP2B, member of RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002886.3 560 Silent Mutation GGC,GGT G26G NP_002877.2

View Full Product Details