Product Details

SNP ID
rs116228921
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:150569483 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGGAGGTCAGGGCTTAATTTCGGC[C/T]AGGTTCTGTTTGAGATGCCCATGAA
Phenotype
MIM: 608087
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GIMAP4 PubMed Links

Gene Details

Gene
GIMAP4
Gene Name
GTPase, IMAP family member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018326.2 Intron NP_060796.1
XM_005250017.1 Intron XP_005250074.1
Gene
LOC107986860
Gene Name
endogenous retrovirus group K member 19 Env polyprotein-like
There are no transcripts associated with this gene.

View Full Product Details