Product Details

SNP ID
rs61734186
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:151349238 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGTTTTTACCACCAAGACTAAAA[A/C]AAGTGAGTAATTTCCCTAAATTTGA
Phenotype
MIM: 607981
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
NUB1 PubMed Links

Gene Details

Gene
NUB1
Gene Name
negative regulator of ubiquitin like proteins 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243351.1 432 Missense Mutation AAA,CAA K119Q NP_001230280.1
NM_016118.4 432 Missense Mutation AAA,CAA K119Q NP_057202.3
XM_005250011.2 432 Missense Mutation AAA,CAA K95Q XP_005250068.1
XM_017012304.1 432 Missense Mutation AAA,CAA K119Q XP_016867793.1
XM_017012305.1 432 Missense Mutation AAA,CAA K119Q XP_016867794.1
XM_017012306.1 432 Missense Mutation AAA,CAA K95Q XP_016867795.1
XM_017012307.1 432 Missense Mutation AAA,CAA K119Q XP_016867796.1
XM_017012308.1 432 Missense Mutation AAA,CAA K119Q XP_016867797.1
XM_017012309.1 432 Missense Mutation AAA,CAA K95Q XP_016867798.1
XM_017012310.1 432 UTR 5 XP_016867799.1
XM_017012311.1 432 UTR 5 XP_016867800.1
XM_017012312.1 432 UTR 5 XP_016867801.1
XM_017012313.1 432 UTR 5 XP_016867802.1
XM_017012314.1 432 UTR 5 XP_016867803.1

View Full Product Details