Product Details

SNP ID
rs62641752
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:141708968 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGTGCATGAGCTCGACACATCTTC[A/G]GAAAAAGACAAAGAAAGTCCAGATC
Phenotype
MIM: 614084
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KIAA1147 PubMed Links

Gene Details

Gene
KIAA1147
Gene Name
KIAA1147
There are no transcripts associated with this gene.

Gene
WEE2
Gene Name
WEE1 homolog 2 (S. pombe)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001105558.1 616 Silent Mutation TCA,TCG S70S NP_001099028.1
Gene
WEE2-AS1
Gene Name
WEE2 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details