Product Details

SNP ID
rs61677869
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:140523495 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCTTGGCCATAGGACACACTGGAG[C/T]CACTGCAGGGCAGGCCTGGCTCAGT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DENND2A PubMed Links

Gene Details

Gene
DENND2A
Gene Name
DENN domain containing 2A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318052.1 Intron NP_001304981.1
NM_001318053.1 Intron NP_001304982.1
NM_015689.4 Intron NP_056504.3
XM_005249977.3 Intron XP_005250034.1
XM_005249979.4 Intron XP_005250036.1
XM_011516052.2 Intron XP_011514354.1
XM_011516053.2 Intron XP_011514355.1
XM_011516054.2 Intron XP_011514356.1
XM_017011989.1 Intron XP_016867478.1

View Full Product Details