Product Details
- SNP ID
-
rs189055696
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:149721503 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGCTCGTGTTCCAGTGGCCCAACTG[G/T]TGACGGGGTCCAGGGAAGTCCTCTC
- Phenotype
-
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
KRBA1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs193130944] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- KRBA1
- Gene Name
- KRAB-A domain containing 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001290187.1 |
756 |
Missense Mutation |
GGT,GTT |
G179V |
NP_001277116.1 |
NM_032534.3 |
756 |
Missense Mutation |
GGT,GTT |
G145V |
NP_115923.2 |
XM_011516636.2 |
756 |
Missense Mutation |
GGT,GTT |
G217V |
XP_011514938.1 |
XM_011516637.2 |
756 |
Missense Mutation |
GGT,GTT |
G216V |
XP_011514939.1 |
XM_011516638.2 |
756 |
Missense Mutation |
GGT,GTT |
G202V |
XP_011514940.1 |
XM_011516639.2 |
756 |
Missense Mutation |
GGT,GTT |
G217V |
XP_011514941.1 |
XM_011516640.2 |
756 |
Missense Mutation |
GGT,GTT |
G216V |
XP_011514942.1 |
XM_011516641.2 |
756 |
Missense Mutation |
GGT,GTT |
G180V |
XP_011514943.1 |
XM_011516642.2 |
756 |
Missense Mutation |
GGT,GTT |
G145V |
XP_011514944.1 |
XM_011516643.2 |
756 |
Missense Mutation |
GGT,GTT |
G145V |
XP_011514945.1 |
View Full Product Details